missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SCYL1BP1 Partial ORF (NP_689494.1, 22 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00092344-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16191207

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 13-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the golgin family, a group of coiled-coil proteins localized to the Golgi. The encoded protein may function in the secretory pathway. The encoded protein, which also localizes to the cytoplasm, was identified by interactions with the N-terminal kinase-like protein, and thus it may function in mitosis. Mutations in this gene have been associated with geroderma osteodysplastica. Alternatively spliced transcript variants have been described. [provided by RefSeq]

Sequence: WPGPMAQGWAGFSEEELRRLKQTKDPFEPQRRLPAKKSRQQLQREKALVEQSQKLGLQDGSTSLLPEQLLSAPKQRVNVQKPPFSSPTLPSHFTLTSPVGDGQPQGIE
Especificaciones

Especificaciones

NP_689494.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WPGPMAQGWAGFSEEELRRLKQTKDPFEPQRRLPAKKSRQQLQREKALVEQSQKLGLQDGSTSLLPEQLLSAPKQRVNVQKPPFSSPTLPSHFTLTSPVGDGQPQGIE
RUO
SCYL1BP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
92344
SCYL1BP1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11752/MGC51263/MGC70512/NTKL-BP1/NTKLBP1/RP11-545I10.1
SCYL1BP1
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales