missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RRM2B Partial ORF (NP_056528, 3 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00050484-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: DPERPEAAGLDQDERSSSDTNESEIKSNEEPLLRKSSRRFVIFPIQYPDIWKMYKQAQASFWTAEEVDLSKDLPHWNKLKADEspecificaciones
NP_056528 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.76kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DPERPEAAGLDQDERSSSDTNESEIKSNEEPLLRKSSRRFVIFPIQYPDIWKMYKQAQASFWTAEEVDLSKDLPHWNKLKAD | |
RUO | |
RRM2B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
50484 | |
RRM2B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686M05248/MGC102856/MGC42116/p53R2 | |
RRM2B | |
Recombinant | |
wheat germ expression system |