Learn More
Abnova™ Human REM1 Partial ORF (NP_054731, 162 a.a. - 251 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00028954-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a GTPase and member of the RAS-like GTP-binding protein family. The encoded protein is expressed in endothelial cells, where it promotes reorganization of the actin cytoskeleton and morphological changes in the cells. [provided by RefSeq]
Sequence: SIADRGSFESASELRIQLRRTHQADHVPIILVGNKADLARCREVSVEEGRACAVVFDCKFIETSATLQHNVAELFEGVVRQLRLRRRDSAEspecificaciones
NP_054731 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SIADRGSFESASELRIQLRRTHQADHVPIILVGNKADLARCREVSVEEGRACAVVFDCKFIETSATLQHNVAELFEGVVRQLRLRRRDSA | |
RUO | |
REM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
28954 | |
REM1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GD:REM/GES/MGC48669/REM | |
REM1 | |
Recombinant | |
wheat germ expression system |