missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human REG1B Partial ORF (NP_006498.1, 67 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005968-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16164235

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 12-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV based on the primary structures of the encoded proteins. This gene encodes a protein secreted by the exocrine pancreas that is highly similar to the REG1A protein. The related REG1A protein is associated with islet cell regeneration and diabetogenesis, and may be involved in pancreatic lithogenesis. Reg family members REG1A, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication. [provided by RefSeq]

Sequence: MNSGNLVSVLTQAEGAFVASLIKESSTDDSNVWIGLHDPKKNRRWHWSSGSLVSYKSWDTGSPSSANAGYCASLTSCSGFKKWKDESCEKKFSFVCKFKN
Especificaciones

Especificaciones

NP_006498.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MNSGNLVSVLTQAEGAFVASLIKESSTDDSNVWIGLHDPKKNRRWHWSSGSLVSYKSWDTGSPSSANAGYCASLTSCSGFKKWKDESCEKKFSFVCKFKN
RUO
REG1B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5968
REG1B (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PSPS2/REGH/REGI-BETA/REGL
REG1B
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales