Learn More
STEMCELL Technologies Human Recombinant IL-2 (E. coli-expressed)
Interleukin 2
Marca:  STEMCELL Technologies 78220
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17159881
  • 99.90€ / 10 microgramos
Fecha estimada de envío: 20-05-2024
para ver el stock
Descripción
Descripción
  • Interleukin 2 (IL-2) is a monomeric cytokine that was originally identified as a T cell growth factor
  • It binds to heterotrimeric receptors consisting of CD25, CD122, and CD132
  • Upon binding, it activates JAK3-, STAT5-, and AKT-dependent signaling pathways, which results in cellular proliferation and survival
  • The majority of IL-2 is secreted by activated CD4+ and CD8+ T cells, although B cells and dendritic cells were found to produce IL-2 in small amounts
  • IL-2 downregulates immune responses to prevent autoimmunity during thymic development, influences the development of CD4+CD25+ T regulatory cells, and affects development of follicular helper T cells
  • IL-2 also controls inflammation by inhibiting Th17 differentiation
  • Human Recombinant IL-2 has a serine substitute for cysteine at position 126
Especificaciones
Especificaciones
Aldesleukin, Interleukin-2, T cell growth factor, TCGF
10 μg
Hematopoietic Stem and Progenitor Cells; NK Cells; T Cells; T Cells, CD4+; T Cells, CD8+
≥95%