Learn More
STEMCELL Technologies Human Recombinant FGF-4
Fibroblast growth factor 4
Marca:  STEMCELL Technologies 78103
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17169571
  • en la oferta / 5 microgramos
Fecha estimada de envío: 24-05-2024
para ver el stock
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
Descripción
Descripción
  • Fibroblast growth factor 4 (FGF-4) is a member of the FGF superfamily (Beenken & Mohammadi)
  • FGF-4 is expressed in pluripotent stem cells and is implicated in various stages of development and morphogenesis in a variety of organisms (Kosaka et al. 2009)
  • FGF-4 has been shown to have an ability to promote neural stem cell proliferation and neuronal differentiation in the postnatal brain (Kosaka et al. 2006)
  • FGF-4 has also been shown to increase the proliferation rate of mesenchymal stem cells (Farré et al.) FGF-4 supports the
  • properties of human embryonic stem cells and also promotes the proliferation of these cells (Mayshar et al.) In the mouse, the Fgf4 gene also supports proliferation of the inner cell mass (ICM) and postimplantation embryos (Feldman et al.)
Especificaciones
Especificaciones
HBGF-4, Heparin-binding growth factor 4, Heparin secretory transforming protein 1, HST, HST-1, HSTF-1, Kaposi sarcoma oncogene, KFGF, Transforming protein KS3
5 μg
Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells
≥95%