Learn More
Abnova™ Human RBPJL Full-length ORF (NP_055091.2, 1 a.a. - 517 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00011317-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
In mouse, recombining binding protein L (RBP-L) is a transcription factor that binds to DNA sequences almost identical to that bound by the Notch receptor signalling pathway transcription factor RBP-J. However, unlike RBP-J, RBP-L does not interact with Notch receptors. RBP-L has been shown to activate transcription in concert with Epstein-Barr virus nuclear antigen-2 (EBNA2). The protein encoded by this gene is similar in sequence to the mouse RPB-L protein and Drosophila suppressor of hairless protein. [provided by RefSeq]
Sequence: MDPAGAADPSVPPNPLTHLSLQDRSEMQLQSEADRRSLPGTWTRSSPEHTTILRGGVRRCLQQQCEQTVRILHAKVAQKSYGNEKRFFCPPPCVYLSGPGWRVKPGQDQAHQAGETGPTVCGYMGLDSASGSATETQKLNFEQQPDSREFGCAKTLYISDADKRKHFRLVLRLVLRGGRELGTFHSRLIKVISKPSQKKQSLKNTDLCISSGSKVSLFNRLRSQTVSTRYLSVEDGAFVASARQWAAFTLHLADGHSAQGDFPPREGYVRYGSLVQLVCTVTGITLPPMIIRKVAKQCALLDVDEPISQLHKCAFQFPGSPPGGGGTYLCLATEKVVQFQASPCPKEANRALLNDSSCWTIIGTESVEFSFSTSLACTLEPVTPVPLISTLELSGGGDVATLELHGENFHAGLKVWFGDVEAETMYRSPRSLVCVVPDVAAFCSDWRWLRAPITIPMSLVRADGLFYPSAFSFTYTPEYSVRPGHPGVPEPATDADALLESIHQEFTRTNFHLFIQTEspecificaciones
NP_055091.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RBP-L/RBPSUHL/SUH/SUHL | |
RBPJL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11317 | |
RBPJL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDPAGAADPSVPPNPLTHLSLQDRSEMQLQSEADRRSLPGTWTRSSPEHTTILRGGVRRCLQQQCEQTVRILHAKVAQKSYGNEKRFFCPPPCVYLSGPGWRVKPGQDQAHQAGETGPTVCGYMGLDSASGSATETQKLNFEQQPDSREFGCAKTLYISDADKRKHFRLVLRLVLRGGRELGTFHSRLIKVISKPSQKKQSLKNTDLCISSGSKVSLFNRLRSQTVSTRYLSVEDGAFVASARQWAAFTLHLADGHSAQGDFPPREGYVRYGSLVQLVCTVTGITLPPMIIRKVAKQCALLDVDEPISQLHKCAFQFPGSPPGGGGTYLCLATEKVVQFQASPCPKEANRALLNDSSCWTIIGTESVEFSFSTSLACTLEPVTPVPLISTLELSGGGDVATLELHGENFHAGLKVWFGDVEAETMYRSPRSLVCVVPDVAAFCSDWRWLRAPITIPMSLVRADGLFYPSAFSFTYTPEYSVRPGHPGVPEPATDADALLESIHQEFTRTNFHLFIQT | |
RUO | |
RBPJL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |