missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Plexin D1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB41601-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Plexin D1 Monoclonal specifically detects Plexin D1 in Human samples. It is validated for Western Blot.
Especificaciones
Plexin D1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q9Y4D7 | |
PLXND1 | |
Mouse myeloma cell line NS0-derived recombinant human Plexin D1 Leu47-Ala1271 Accession # Q9Y4D7 | |
25 μg | |
Primary | |
Detects human Plexin D1 in ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
752815 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1, PLXND1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23129 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human Plexin D1 Antibody, R&D Systems™ > 25μg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido