Learn More
Abnova™ Human PLAGL1 Partial ORF (NP_002647.2, 221 a.a. - 320 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005325-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a C2H2 zinc finger protein with transactivation and DNA-binding activity. This gene has been shown to exhibit antiproliferative activities and is a tumor suppressor gene candidate. Many transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]
Sequence: QPMQPLPESLASLHPSVSPGSPPPPLPNHKYNTTSTSYSPLASLPLKADTKGFCNISLFEDLPLQEPQSPQKLNPGFDLAKGNAGKVNLPKELPADAVNLEspecificaciones
NP_002647.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QPMQPLPESLASLHPSVSPGSPPPPLPNHKYNTTSTSYSPLASLPLKADTKGFCNISLFEDLPLQEPQSPQKLNPGFDLAKGNAGKVNLPKELPADAVNL | |
RUO | |
PLAGL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5325 | |
PLAGL1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781P1017/LOT1/MGC126275/MGC126276/ZAC/ZAC1 | |
PLAGL1 | |
Recombinant | |
wheat germ expression system |