missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PIGZ Full-length ORF (AAH44640.1, 1 a.a. - 579 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00080235-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16124873

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 13-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The glycosylphosphatidylinositol (GPI) anchor is a glycolipid found on many blood cells that serves to anchor proteins to the cell surface. This gene encodes a protein that is localized to the endoplasmic reticulum, and is involved in GPI anchor biosynthesis. As shown for the yeast homolog, which is a member of a family of dolichol-phosphate-mannose (Dol-P-Man)-dependent mannosyltransferases, this protein can also add a side-branching fourth mannose to GPI precursors during the assembly of GPI anchors. [provided by RefSeq]

Sequence: MQICGSSVASVAAGTSFQVLGPVCWQQLDLKMAVRVLWGGLSLLRVLWCLLPQTGYVHPDEFFQSPEVMAEDILGVQAARPWEFYPSSSCRSVLFPLLISGSTFWLLRLWEELGPWPGLVSGYALLVGPRLLLTALSFALDGAVYHLAPPMGADRWNALALLSGSYVTLVFYTRTFSNTIEGLLFTWLLVLVSSHVTWGPTRKEPAPGPRWRSWLLGGIVAAGFFNRPTFLAFAVVPLYLWGTRGATNPGLKSLTREALVLLPGATLTAAVFVATDSWYFSSPATSRNLVLTPVNFLHYNLNPQNLARHGTHARLTHLAVNGFLLFGVLHAQALQAAWQQLQVGLQASAQMGLLRALGARSLLSSPRSYLLLLYFMPLALLSAFSHQEARFLIPLLVPLVLLCSPQTQPVPWKGTVVLFNALGALLFGCLHQGGLVPGLEYLEQVVHAPVLPSTPTHYTLLFTHTYMPPRHLLHLPGLGAPVEVVDIGGTEDWALCQTLKSFTRQPACQVAGGPWLCRLFVVTPGTTRRAVEKCSFPFKNETLLFPHLTLEDPPALSSLLSGAWRDHLSLHIVELGEET
Especificaciones

Especificaciones

AAH44640.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12768/GPI-MT-IV/MGC52163/SMP3
PIGZ
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
80235
PIGZ (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQICGSSVASVAAGTSFQVLGPVCWQQLDLKMAVRVLWGGLSLLRVLWCLLPQTGYVHPDEFFQSPEVMAEDILGVQAARPWEFYPSSSCRSVLFPLLISGSTFWLLRLWEELGPWPGLVSGYALLVGPRLLLTALSFALDGAVYHLAPPMGADRWNALALLSGSYVTLVFYTRTFSNTIEGLLFTWLLVLVSSHVTWGPTRKEPAPGPRWRSWLLGGIVAAGFFNRPTFLAFAVVPLYLWGTRGATNPGLKSLTREALVLLPGATLTAAVFVATDSWYFSSPATSRNLVLTPVNFLHYNLNPQNLARHGTHARLTHLAVNGFLLFGVLHAQALQAAWQQLQVGLQASAQMGLLRALGARSLLSSPRSYLLLLYFMPLALLSAFSHQEARFLIPLLVPLVLLCSPQTQPVPWKGTVVLFNALGALLFGCLHQGGLVPGLEYLEQVVHAPVLPSTPTHYTLLFTHTYMPPRHLLHLPGLGAPVEVVDIGGTEDWALCQTLKSFTRQPACQVAGGPWLCRLFVVTPGTTRRAVEKCSFPFKNETLLFPHLTLEDPPALSSLLSGAWRDHLSLHIVELGEET
RUO
PIGZ
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales