Learn More
Abnova™ Human PIGZ Full-length ORF (AAH44640.1, 1 a.a. - 579 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080235-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The glycosylphosphatidylinositol (GPI) anchor is a glycolipid found on many blood cells that serves to anchor proteins to the cell surface. This gene encodes a protein that is localized to the endoplasmic reticulum, and is involved in GPI anchor biosynthesis. As shown for the yeast homolog, which is a member of a family of dolichol-phosphate-mannose (Dol-P-Man)-dependent mannosyltransferases, this protein can also add a side-branching fourth mannose to GPI precursors during the assembly of GPI anchors. [provided by RefSeq]
Sequence: MQICGSSVASVAAGTSFQVLGPVCWQQLDLKMAVRVLWGGLSLLRVLWCLLPQTGYVHPDEFFQSPEVMAEDILGVQAARPWEFYPSSSCRSVLFPLLISGSTFWLLRLWEELGPWPGLVSGYALLVGPRLLLTALSFALDGAVYHLAPPMGADRWNALALLSGSYVTLVFYTRTFSNTIEGLLFTWLLVLVSSHVTWGPTRKEPAPGPRWRSWLLGGIVAAGFFNRPTFLAFAVVPLYLWGTRGATNPGLKSLTREALVLLPGATLTAAVFVATDSWYFSSPATSRNLVLTPVNFLHYNLNPQNLARHGTHARLTHLAVNGFLLFGVLHAQALQAAWQQLQVGLQASAQMGLLRALGARSLLSSPRSYLLLLYFMPLALLSAFSHQEARFLIPLLVPLVLLCSPQTQPVPWKGTVVLFNALGALLFGCLHQGGLVPGLEYLEQVVHAPVLPSTPTHYTLLFTHTYMPPRHLLHLPGLGAPVEVVDIGGTEDWALCQTLKSFTRQPACQVAGGPWLCRLFVVTPGTTRRAVEKCSFPFKNETLLFPHLTLEDPPALSSLLSGAWRDHLSLHIVELGEETEspecificaciones
AAH44640.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12768/GPI-MT-IV/MGC52163/SMP3 | |
PIGZ | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80235 | |
PIGZ (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQICGSSVASVAAGTSFQVLGPVCWQQLDLKMAVRVLWGGLSLLRVLWCLLPQTGYVHPDEFFQSPEVMAEDILGVQAARPWEFYPSSSCRSVLFPLLISGSTFWLLRLWEELGPWPGLVSGYALLVGPRLLLTALSFALDGAVYHLAPPMGADRWNALALLSGSYVTLVFYTRTFSNTIEGLLFTWLLVLVSSHVTWGPTRKEPAPGPRWRSWLLGGIVAAGFFNRPTFLAFAVVPLYLWGTRGATNPGLKSLTREALVLLPGATLTAAVFVATDSWYFSSPATSRNLVLTPVNFLHYNLNPQNLARHGTHARLTHLAVNGFLLFGVLHAQALQAAWQQLQVGLQASAQMGLLRALGARSLLSSPRSYLLLLYFMPLALLSAFSHQEARFLIPLLVPLVLLCSPQTQPVPWKGTVVLFNALGALLFGCLHQGGLVPGLEYLEQVVHAPVLPSTPTHYTLLFTHTYMPPRHLLHLPGLGAPVEVVDIGGTEDWALCQTLKSFTRQPACQVAGGPWLCRLFVVTPGTTRRAVEKCSFPFKNETLLFPHLTLEDPPALSSLLSGAWRDHLSLHIVELGEET | |
RUO | |
PIGZ | |
Wheat Germ (in vitro) | |
GST | |
Liquid |