Learn More
Abnova™ Human PCLO Full-length ORF (AAH01304.2, 1 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027445-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Synaptic vesicles dock and fuse in the active zone of the plasma membrane at chemical synapses. The presynaptic cytoskeletal matrix (PCM), which is associated with the active zone and is situated between synaptic vesicles, is thought to be involved in maintaining the neurotransmitter release site in register with the postsynaptic reception apparatus. The cycling of synaptic vesicles is a multistep process involving a number of proteins (see MIM 603215). Among the components of the PCM that orchestrate these events are Bassoon (BSN; MIM 604020), RIM (RBBP8; MIM 604124), Oboe, and Piccolo (PCLO).[supplied by OMIM]
Sequence: MKKFRVSLVSKVGKQKYVDLNMLSDSENSQHLELHEPPKAVDKAKSPGVDPKQLAAELQKVSLQQSPLVLSSVVEKGSHVHSGPTSAGSSSVPSPGQPGSPSVSKKKHGSSKPTDGTKVVSHPITGEIQLQINYDLGNLIIHILQARNLVPRDNNGYSDPFVKVYLLPGRGAEYKRRTKHVQKSLNPEWNQTVIYKSISMEQLKKKTLEVTVWDYDRFSSNDFLGEVLIDLSSTAHLDNTPRWYPLKEQTESIDHGKSHSSQSSQQSPKPSVIKSRSHGIFPDPSKDMQVPTIEKSHSSPGSSKSSSEGHLRSHGPSRSQSKTSVTQTHLEDAGAAIAAAEAAVQQLRIQPSKRRKEspecificaciones
AAH01304.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACZ/DKFZp779G1236 | |
PCLO | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27445 | |
PCLO (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKKFRVSLVSKVGKQKYVDLNMLSDSENSQHLELHEPPKAVDKAKSPGVDPKQLAAELQKVSLQQSPLVLSSVVEKGSHVHSGPTSAGSSSVPSPGQPGSPSVSKKKHGSSKPTDGTKVVSHPITGEIQLQINYDLGNLIIHILQARNLVPRDNNGYSDPFVKVYLLPGRGAEYKRRTKHVQKSLNPEWNQTVIYKSISMEQLKKKTLEVTVWDYDRFSSNDFLGEVLIDLSSTAHLDNTPRWYPLKEQTESIDHGKSHSSQSSQQSPKPSVIKSRSHGIFPDPSKDMQVPTIEKSHSSPGSSKSSSEGHLRSHGPSRSQSKTSVTQTHLEDAGAAIAAAEAAVQQLRIQPSKRRK | |
RUO | |
PCLO | |
Wheat Germ (in vitro) | |
GST | |
Liquid |