Learn More
Abnova™ Human NUP37 Full-length ORF (NP_076962.2, 1 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00079023-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP37 is part of one such subcomplex, Nup107-160 (Loiodice et al., 2004 [PubMed 15146057]).[supplied by OMIM]
Sequence: MKQDASRNAAYTVDCEDYVHVVEFNPFENGDSGNLIAYGGNNYVVIGTCTFQEEEADVEGIQYKTLRTFHHGVRVDGIAWSPETRLDSLPPVIKFCTSAADMKIRLFTSDLQDKNEYKVLEGHTDFINGLVFDPKEGQEIASVSDDHTCRIWNLEGVQTAHFVLHSPGMSVCWHPEETFKLMVAEKNGTIRFYDLLAQQAILSLESEQVPLMSAHWCLKNTFKVGAVAGNDWLIWDITRSSYPQNKRPVHMDRACLFRWSTISENLFATTGYPGKMASQFQIHHLGHPQPILMGSVAVGSGLSWHRTLPLCVIGGDHKLLFWVTEVEspecificaciones
NP_076962.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22618/MGC5585/p37 | |
NUP37 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79023 | |
NUP37 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKQDASRNAAYTVDCEDYVHVVEFNPFENGDSGNLIAYGGNNYVVIGTCTFQEEEADVEGIQYKTLRTFHHGVRVDGIAWSPETRLDSLPPVIKFCTSAADMKIRLFTSDLQDKNEYKVLEGHTDFINGLVFDPKEGQEIASVSDDHTCRIWNLEGVQTAHFVLHSPGMSVCWHPEETFKLMVAEKNGTIRFYDLLAQQAILSLESEQVPLMSAHWCLKNTFKVGAVAGNDWLIWDITRSSYPQNKRPVHMDRACLFRWSTISENLFATTGYPGKMASQFQIHHLGHPQPILMGSVAVGSGLSWHRTLPLCVIGGDHKLLFWVTEV | |
RUO | |
NUP37 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |