Learn More
Abnova™ Human NT5DC2 Full-length ORF (NP_075059.1, 1 a.a. - 520 a.a.) Recombinant Protein with GST-tag at N-terminal
Recombinant proteins expressed from in vitro wheat germ system.
Marca: Abnova™ H00064943-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MRVESGSAQERGILLESLSTLLEKTTASHEGRAPGNRELTDLLPPEVCSLLNPAAIYANNEISLRDVEVYGFDYDYTLAQYADALHPEIFSTARDILIEHYKYPEGIRKYDYNPSFAIRGLHYDIQKSLLMKIDAFHYVQLGTAYR
Especificaciones
NP_075059.1 | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
NT5DC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MRVESGSAQERGILLESLSTLLEKTTASHEGRAPGNRELTDLLPPEVCSLLNPAAIYANNEISLRDVEVYGFDYDYTLAQYADALHPEIFSTARDILIEHYKYPEGIRKYDYNPSFAIRGLHYDIQKSLLMKIDAFHYVQLGTAYRGLQPVPDEEVIELYGGTQHIPLYQMSGFYGKGPSIKQFMDIFSLPEMALLSCVVDYFLGHSLEFDQAHLYKDVTDAIRDVHVKGLMYQWIEQDMEKYILRGDETFAVLSRLVAHGKQLFLITNSPFSFVDKGMRHMVGPDWRQLFDVVIVQADKPSFFTDRRKPFRKLDEKGSLQWDRITRLEKGKIYRQGNLFDFLRLTEWRGPRVLYFGDHLYSDLADLMLRHGWRTGAIIPELEREIRIINTEQYMHSLTWQQALTGLLERMQTYQDAESRQVLAAWMKERQELRCITKALFNAQFGSIFRTFHNPTYFSRRLVRFSDLYMASLSCLLNYRVDFTFYPRRTPLQHEAPLWMDQLCTGCMKTPFLGDMAHIR | |
RUO | |
NT5DC2 | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
64943 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12442 | |
NT5DC2 | |
Liquid |
For Research Use Only