Learn More
Abnova™ Human NRF1 Full-length ORF (AAH16925.1, 1 a.a. - 522 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for nuclear factor (erythroid-derived 2)-like 1 which has an official symbol of NFE2L1. [provided by RefSeq]
Especificaciones
Especificaciones
Número de acceso | AAH16925.1 |
Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 4899 |
Peso molecular | 82.1kDa |
Nombre | NRF1 (Human) Recombinant Protein (P01) |
Método de purificación | Glutathione Sepharose 4 Fast Flow |
Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Cantidad | 25 ug |
Inmunógeno | MEEHGVTQTEHMATIEAHAVAQQVQQVHVATYTEHSMLSADEDSPSSPEDTSYDDSDILNSTAADEVTAHLAAAGPVGMAAAAAVATGKKRKRPHVFESNPSIRKRQQTRLLRKLRATLDEYTTRVGQQAIVLCISPSKPNPVFKVFGAAPLENVVRKYKSMILEDLESALAEHAPAPQEVNSELPPLTIDGIPVSVDKMTQAQLRAFIPEMLKYSTGRGKPGWGKESCKPIWWPEDIPWANVRSDVRTEEQKQRVSWTQALRTIVKNCYKQHGREDLLYAFEDQQTQTQATATHSIAHLVPSQTVVQTFSNPDGTVSLIQVGTGATVATLADASELPTTVTVAQVNYSAVADGEVEQNWATLQGGEMTIQTTQASEATQAVASLAEAAVAASQEMQQGATVTMALNSEAAAHAVATLAEATLQGGGQIVLSGETAAAVGALTGVQDANGLFMADRAGRKWILTDKATGLVQIPVSMYQTVVTSLAQGNGPVQVAMAPVTTRISDSAVTMDGQAVEVVTLEQ |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.