missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human NKG2A&CD94 Protein, Mouse IgG2a Fc Tag
423.38€ - 3108.82€
Especificaciones
Número de acceso | NP_002250.1 (NKG2A) and N/A |
---|---|
Peso molecular | 55.5 kDa |
Fuente | Human NKG2A&CD94;, Mouse IgG2a Fc Tag (NC4-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) and Asp 57 - Ile 179 (CD94) (Accession # P26715-1 (NKG2A) and Q13241-1 (CD94)). |
Inmunógeno | Ala 113 - Leu 233 (NKG2A) and Asp 57 - Ile 179 (CD94) |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17176521
|
ACROBiosystems
NC4-H5253-100UG |
100 μg |
423.38€
100 microgramos |
Fecha estimada de envío: 07-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
17186521
|
ACROBiosystems
NC4-H5253-1MG |
1 mg |
3108.82€
1 miligramo |
Fecha estimada de envío: 07-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. NKG2A/CD159a is a transmembrane protein belonging to the CD94/NKG2 family of C-type lectin-like receptors that inhibits innate immune system activation. CD94 pairs with the NKG2 molecule as a heterodimer. The CD94/NKG2 complex, on the surface of natural killer cells interacts with Human Leukocyte Antigen (HLA)-E on target cells.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 55.5 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_002250.1 (NKG2A) and N/A | |
Human NKG2A&CD94;, Mouse IgG2a Fc Tag (NC4-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) and Asp 57 - Ile 179 (CD94) (Accession # P26715-1 (NKG2A) and Q13241-1 (CD94)). | |
-20°C | |
NKG2A and CD94 | |
Human | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
55.5 kDa | |
Ala 113 - Leu 233 (NKG2A) and Asp 57 - Ile 179 (CD94) | |
NKG2A and CD94 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |