Learn More
Abnova™ Human NFKB1 Partial ORF (AAH51765, 860 a.a. - 969 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
This gene encodes a 105 kD protein which can undergo cotranslational processing by the 26S proteasome to produce a 50 kD protein. The 105 kD protein is a Rel protein-specific transcription inhibitor and the 50 kD protein is a DNA binding subunit of the NF-kappa-B (NFKB) protein complex. NFKB is a transcription regulator that is activated by various intra- and extra-cellular stimuli such as cytokines, oxidant-free radicals, ultraviolet irradiation, and bacterial or viral products. Activated NFKB translocates into the nucleus and stimulates the expression of genes involved in a wide variety of biological functions. Inappropriate activation of NFKB has been associated with a number of inflammatory diseases while persistent inhibition of NFKB leads to inappropriate immune cell development or delayed cell growth. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Especificaciones
Especificaciones
Número de acceso | AAH51765 |
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 4790 |
Peso molecular | 37.73kDa |
Nombre | NFKB1 (Human) Recombinant Protein (Q01) |
Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Cantidad | 25 ug |
Inmunógeno | MDNYEVSGGTVRELVEALRQMGYTEAIEVIQAASSPVKTTSQAHSLPLSPASTRQQIDELRDSDSVCDSGVETSFRKLSFTESLTSGASLLTLNKMPHDYGQEGPLEGKI |
Requisitos de almacenamiento | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.