Learn More
Abnova™ Human NCOA1 Partial ORF (NP_003734, 284 a.a. - 393 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008648-Q01L
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene acts as a transcriptional coactivator for steroid and nuclear hormone receptors. It is a member of the p160/steroid receptor coactivator (SRC) family and like other family members has histone acetyltransferase activity and contains a nuclear localization signal, as well as bHLH and PAS domains. The product of this gene binds nuclear receptors directly and stimulates the transcriptional activities in a hormone-dependent fashion. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: GRTGWEDLVRKCIYAFFQPQGREPSYARQLFQEVMTRGTASSPSYRFILNDGTMLSAHTKCKLCYPQSPDMQPFIMGIHIIDREHSGLSPQDDTNSGMSIPRVNPSVNPSEspecificaciones
NP_003734 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GRTGWEDLVRKCIYAFFQPQGREPSYARQLFQEVMTRGTASSPSYRFILNDGTMLSAHTKCKLCYPQSPDMQPFIMGIHIIDREHSGLSPQDDTNSGMSIPRVNPSVNPS | |
RUO | |
NCOA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8648 | |
NCOA1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
F-SRC-1/KAT13A/MGC129719/MGC129720/NCoA-1/RIP160/SRC-1/SRC1/bHLHe42/bHLHe74 | |
NCOA1 | |
Recombinant | |
wheat germ expression system |