Learn More
Abnova™ Human MXD1 Full-length ORF (NP_002348.1, 1 a.a. - 221 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004084-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor. [provided by RefSeq]
Sequence: MAAAVRMNIQMLLEAADYLERREREAEHGYASMLPYNNKDRDALKRRNKSKKNNSSSRSTHNEMEKNRRAHLRLCLEKLKGLVPLGPESSRHTTLSLLTKAKLHIKKLEDCDRKAVHQIDQLQREQRHLKRQLEKLGIERIRMDSIGSTVSSERSDSDREEIDVDVESTDYLTGDLDWSSSSVSDSDERGSMQSLGSDEGYSSTSIKRIKLQDSHKACLGLEspecificaciones
NP_002348.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAD/MAD1/MGC104659 | |
MXD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4084 | |
MXD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAAVRMNIQMLLEAADYLERREREAEHGYASMLPYNNKDRDALKRRNKSKKNNSSSRSTHNEMEKNRRAHLRLCLEKLKGLVPLGPESSRHTTLSLLTKAKLHIKKLEDCDRKAVHQIDQLQREQRHLKRQLEKLGIERIRMDSIGSTVSSERSDSDREEIDVDVESTDYLTGDLDWSSSSVSDSDERGSMQSLGSDEGYSSTSIKRIKLQDSHKACLGL | |
RUO | |
MXD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |