missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MVK Partial ORF (AAH16140, 297 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004598-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108041

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 12-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes the peroxisomal enzyme mevalonate kinase. Mevalonate is a key intermediate, and mevalonate kinase a key early enzyme, in isoprenoid and sterol synthesis. Mevalonate kinase deficiency caused by mutation of this gene results in mevalonic aciduria, a disease characterized psychomotor retardation, failure to thrive, hepatosplenomegaly, anemia and recurrent febrile crises. Defects in this gene also cause hyperimmunoglobulinaemia D and periodic fever syndrome, a disorder characterized by recurrent episodes of fever associated with lymphadenopathy, arthralgia, gastrointestinal dismay and skin rash. Two transcript variants that encode the same protein have been found for this gene. [provided by RefSeq]

Sequence: LIDMNQHHLNALGVGHASLDQLCQVTRARGLHSKLTGAGGGGCGITLLKPGLEQPEVEATKQALTSCGFDCLETSIGAPGVSIHSATSLDSRVQQALDGL
Especificaciones

Especificaciones

AAH16140
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LIDMNQHHLNALGVGHASLDQLCQVTRARGLHSKLTGAGGGGCGITLLKPGLEQPEVEATKQALTSCGFDCLETSIGAPGVSIHSATSLDSRVQQALDGL
RUO
MVK
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4598
MVK (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ96772/LRBP/MK/MVLK
MVK
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales