missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Smad7 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB2029-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Smad7 Monoclonal specifically detects Smad7 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Especificaciones
Smad7 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL | |
O15105 | |
SMAD7 | |
E. coli-derived recombinant human Smad7 Gly320-Ser398 Accession # O15105 | |
25 μg | |
Primary | |
Detects human, mouse, and rat Smad7. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
293039 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7, MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4092 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human/Mouse/Rat Smad7 Antibody, R&D Systems™ > 25μg; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido