missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat BAT3/BAG6 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marca: R&D Systems AF6438
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BAT3/BAG6 Polyclonal specifically detects BAT3/BAG6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Especificaciones
BAT3/BAG6 | |
Polyclonal | |
Unconjugated | |
P46379 | |
BAG6 | |
E. coli-derived recombinant human BAT3/BAG6 Met1-Gln190 Accession # P46379 | |
100 μg | |
Primary | |
Detects human, mouse, and rat BAT3/BAG6 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
BAG6, BAT3HLA-B associated transcript-3, BCL2-associated athanogene 6BAG-6, D6S52E, D6S52EHLA-B-associated transcript 3, G3BAG family molecular chaperone regulator 6, HLA-B associated transcript 3, large proline-rich protein BAT3, Protein G3, Protein Scythe, scythe | |
Sheep | |
Affinity Purified | |
RUO | |
7917 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human/Mouse/Rat BAT3/BAG6 Antibody, R&D Systems™ > 100μg; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido