Learn More
Abnova™ Human MAGED4B Full-length ORF (AAH01207.1, 1 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00081557-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the MAGED gene family. It is expressed only in brain and ovary among normal tissues, and two transcript variants of this gene are specifically expressed in glioma cells among cancer cells. This gene and the other MAGED genes are clustered on chromosome Xp11. Multiple alternatively spliced transcript variants have been found for this gene, however, the full-length nature of some variants has not been defined. [provided by RefSeq]
Sequence: MIKDYKKIPIKRADMLKDVIREYDEHFPEIIERATYTLEKKFGIHLKEIDKEEHLYILVCTRDSSARLLGKTKDTPRLSLLLVILGVIFMNGNRASEAVLWEALRKMGLRPGVRHPFLGDLRKLITDDFVKQKNPELREETPLSMAPSWYLEYKKIPNSNPPEYEFLWGLRARHETSKMRVLRFIAQNQNRDPREWKAHFLEAVDDAFKTMDVDMAEEHARAQMRAQMNIGDEALIGRWSWDDIQVELLTWDEDGDFGDAWARIPFAFWARYHQYILNSNRANRRATWRAGVSSGTNGGASTSVLDGPSTSSTIRTRNAARAGASFFSWIQHREspecificaciones
AAH01207.1 | |
Liquid | |
81557 | |
MAGED4B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIKDYKKIPIKRADMLKDVIREYDEHFPEIIERATYTLEKKFGIHLKEIDKEEHLYILVCTRDSSARLLGKTKDTPRLSLLLVILGVIFMNGNRASEAVLWEALRKMGLRPGVRHPFLGDLRKLITDDFVKQKNPELREETPLSMAPSWYLEYKKIPNSNPPEYEFLWGLRARHETSKMRVLRFIAQNQNRDPREWKAHFLEAVDDAFKTMDVDMAEEHARAQMRAQMNIGDEALIGRWSWDDIQVELLTWDEDGDFGDAWARIPFAFWARYHQYILNSNRANRRATWRAGVSSGTNGGASTSVLDGPSTSSTIRTRNAARAGASFFSWIQHR | |
RUO | |
MAGED4B | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC3210/MGC88639 | |
MAGED4B | |
Yes | |
wheat germ expression system |