Learn More
ACROBiosystems Human MAG / Siglec-4a Protein, Fc Tag
Marca:  ACROBiosystems MAG-H5254-1mg
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16477859
  • 2639.24€ / 1 miligramo
Fecha estimada de envío: 11-04-2024
para ver el stock
Descripción
Descripción
Myelin-associated glycoprotein (MAG), a nervous system cell adhesion molecule, is an I-type lectin that binds to sialylated glycoconjugates, including gangliosides bearing characteristic structural determinants. Preferentially binds to alpha-2,3-linked sialic acid. Binds ganglioside Gt1b. Adhesion molecule that mediates interactions between myelinating cells and neurons by binding to neuronal sialic acid-containing gangliosides and to the glycoproteins RTN4R and RTN4RL2. Protection against apoptosis is probably mediated via interaction with neuronal RTN4R and RTN4RL2. In dorsal root ganglion neurons the inhibition is mediated primarily via binding to neuronal RTN4R or RTN4RL2 and to a lesser degree via binding to neuronal gangliosides. In cerebellar granule cells the inhibition is mediated primarily via binding to neuronal gangliosides. In sensory neurons, inhibition of neurite extension depends only partially on RTN4R, RTN4RL2 and gangliosides.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.1 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
NP_002352.1
1 mg
Gly 20 - Pro 516
MAG,Siglec-4 A,GMA,S-MAG
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
81.1 kDa
Human MAG, Fc Tag (MAG-H5254) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Pro 516 (Accession # P20916-1).
-20°C
MAG / Siglec-4 A
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens