Learn More
Abnova™ Human MAB21L1 Partial ORF (NP_005575, 250 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004081-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is similar to the MAB-21 cell fate-determining gene found in C. elegans. It may be involved in eye and cerebellum development, and it has been proposed that expansion of a trinucleotide repeat region in the 5' UTR may play a role in a variety of psychiatric disorders. [provided by RefSeq]
Sequence: CLSILKTLRDRHLELPGQPLNNYHMKTLVSYECEKHPRESDWDESCLGDRLNGILLQLISCLQCRRCPHYFLPNLDLFQGKPHSALENAAKQTWRLAREILTNPKSLEKEspecificaciones
NP_005575 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CLSILKTLRDRHLELPGQPLNNYHMKTLVSYECEKHPRESDWDESCLGDRLNGILLQLISCLQCRRCPHYFLPNLDLFQGKPHSALENAAKQTWRLAREILTNPKSLEK | |
RUO | |
MAB21L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4081 | |
MAB21L1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAGR1/FLJ10197/Nbla00126 | |
MAB21L1 | |
Recombinant | |
wheat germ expression system |