Learn More
Abnova™ Human KRT75 Full-length ORF (AAI60175.1, 1 a.a. - 551 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009119-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. This gene is expressed in the companion layer, upper germinative matrix region of the hair follicle, and medulla of the hair shaft. The encoded protein plays an essential role in hair and nail formation. Variations in this gene have been associated with the hair disorders pseudofolliculitis barbae (PFB) and loose anagen hair syndrome (LAHS). [provided by RefSeq]
Sequence: MSRQSSITFQSGSRRGFSTTSAITPAAGRSRFSSVSVARSAAGSGGLGRISSAGASFGSRSLYNLGGAKRVSINGCGSSCRSGFGGRASNRFGVNSGFGYGGGVGGGFSGPSFPVCPPGGIQEVTVNQSLLTPLHLQIDPTIQRVRAEEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGSRTVRQNLEPLFDSYTSELRRQLESITTERGRLEAELRNMQDVVEDFKVRYEDEINKRTAAENEFVALKKDVDAAYMNKVELEAKVKSLPEEINFIHSVFDAELSQLQTQVGDTSVVLSMDNNRNLDLDSIIAEVKAQYEDIANRSRAEAESWYQTKYEELQVTAGRHGDDLRNTKQEISEMNRMIQRLRAEIDSVKKQCSSLQTAIADAEQRGELALKDARAKLVDLEEALQKAKQDMARLLREYQELMNIKLALDVEIATYRKLLEGEECRLSGEGVSPVNISVVTSTLSSGYGSGSSIGGGNLGLGGGSGYSFTTSGGHSLGAGLGGSGFSATSNRGLGGSGSSVKFVSTTSSSQKSYTHEspecificaciones
AAI60175.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
K6HF/KB18/PFB | |
KRT75 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9119 | |
KRT75 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MSRQSSITFQSGSRRGFSTTSAITPAAGRSRFSSVSVARSAAGSGGLGRISSAGASFGSRSLYNLGGAKRVSINGCGSSCRSGFGGRASNRFGVNSGFGYGGGVGGGFSGPSFPVCPPGGIQEVTVNQSLLTPLHLQIDPTIQRVRAEEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGSRTVRQNLEPLFDSYTSELRRQLESITTERGRLEAELRNMQDVVEDFKVRYEDEINKRTAAENEFVALKKDVDAAYMNKVELEAKVKSLPEEINFIHSVFDAELSQLQTQVGDTSVVLSMDNNRNLDLDSIIAEVKAQYEDIANRSRAEAESWYQTKYEELQVTAGRHGDDLRNTKQEISEMNRMIQRLRAEIDSVKKQCSSLQTAIADAEQRGELALKDARAKLVDLEEALQKAKQDMARLLREYQELMNIKLALDVEIATYRKLLEGEECRLSGEGVSPVNISVVTSTLSSGYGSGSSIGGGNLGLGGGSGYSFTTSGGHSLGAGLGGSGFSATSNRGLGGSGSSVKFVSTTSSSQKSYTH | |
RUO | |
KRT75 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |