missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human JARID2 Partial ORF (NP_004964, 1130 a.a. - 1229 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003720-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16170345

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 28-04-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is an ortholog of the mouse jumonji gene, which encodes a nuclear protein essential for mouse embryogenesis, including neural tube formation. Overexpression of mouse jumonji negatively regulates cell proliferation. The jumonji proteins contain a DNA-binding domain, called an AT-rich interaction domain (ARID), and share regions of similarity with human retinoblastoma-binding protein-2 and the human SMCX protein. [provided by RefSeq]

Sequence: LQLETSERRCQICQHLCYLSMVVQENENVVFCLECALRHVEKQKSCRGLKLMYRYDEEQIISLVNQICGKVSGKNGSIENCLSKPTPKRGPRKRATVDVP
Especificaciones

Especificaciones

NP_004964
Liquid
3720
JARID2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
JMJ
JARID2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LQLETSERRCQICQHLCYLSMVVQENENVVFCLECALRHVEKQKSCRGLKLMYRYDEEQIISLVNQICGKVSGKNGSIENCLSKPTPKRGPRKRATVDVP
RUO
JARID2
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales