Learn More
Abnova™ Human ITPKC Full-length ORF (NP_079470.1, 1 a.a. - 683 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080271-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the inositol 1,4,5-trisphosphate [Ins(1,4,5)P(3)] 3-kinase family of enzymes that catalyze the phosphorylation of inositol 1,4,5-trisphosphate to 1,3,4,5-tetrakisphosphate. The encoded protein is localized to the nucleus and cytoplasm and has both nuclear import and nuclear export activity. Single nucleotide polymorphisms in this gene are associated with Kawasaki disease
Sequence: MRRCPCRGSLNEAEAGALPAAARMGLEAPRGGRRRQPGQQRPGPGAGAPAGRPEGGGPWARTEGSSLHSEPERAGLGPAPGTESPQAEFWTDGQTEPAAAGLGVETERPKQKTEPDRSSLRTHLEWSWSELETTCLWTETGTDGLWTDPHRSDLQFQPEEASPWTQPGVHGPWTELETHGSQTQPERVKSWADNLWTHQNSSSLQTHPEGACPSKEPSADGSWKELYTDGSRTQQDIEGPWTEPYTDGSQKKQDTEAARKQPGTGGFQIQQDTDGSWTQPSTDGSQTAPGTDCLLGEPEDGPLEEPEPGELLTHLYSHLKCSPLCPVPRLIITPETPEPEAQPVGPPSRVEGGSGGFSSASSFDESEDDVVAGGGGASDPEDRSGSKPWKKLKTVLKYSPFVVSFRKHYPWVQLSGHAGNFQAGEDGRILKRFCQCEQRSLEQLMKDPLRPFVPAYYGMVLQDGQTFNQMEDLLADFEGPSIMDCKMGSRTYLEEELVKARERPRPRKDMYEKMVAVDPGAPTPEEHAQGAVTKPRYMQWRETMSSTSTLGFRIEGIKKADGTCNTNFKKTQALEQVTKVLEDFVDGDHVILQKYVACLEELREALEISPFFKTHEVVGSSLLFVHDHTGLAKVWMIDFGKTVALPDHQTLSHRLPWAEGNREDGYLWGLDNMICLLQGLAQSEspecificaciones
NP_079470.1 | |
Liquid | |
80271 | |
ITPKC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRRCPCRGSLNEAEAGALPAAARMGLEAPRGGRRRQPGQQRPGPGAGAPAGRPEGGGPWARTEGSSLHSEPERAGLGPAPGTESPQAEFWTDGQTEPAAAGLGVETERPKQKTEPDRSSLRTHLEWSWSELETTCLWTETGTDGLWTDPHRSDLQFQPEEASPWTQPGVHGPWTELETHGSQTQPERVKSWADNLWTHQNSSSLQTHPEGACPSKEPSADGSWKELYTDGSRTQQDIEGPWTEPYTDGSQKKQDTEAARKQPGTGGFQIQQDTDGSWTQPSTDGSQTAPGTDCLLGEPEDGPLEEPEPGELLTHLYSHLKCSPLCPVPRLIITPETPEPEAQPVGPPSRVEGGSGGFSSASSFDESEDDVVAGGGGASDPEDRSGSKPWKKLKTVLKYSPFVVSFRKHYPWVQLSGHAGNFQAGEDGRILKRFCQCEQRSLEQLMKDPLRPFVPAYYGMVLQDGQTFNQMEDLLADFEGPSIMDCKMGSRTYLEEELVKARERPRPRKDMYEKMVAVDPGAPTPEEHAQGAVTKPRYMQWRETMSSTSTLGFRIEGIKKADGTCNTNFKKTQALEQVTKVLEDFVDGDHVILQKYVACLEELREALEISPFFKTHEVVGSSLLFVHDHTGLAKVWMIDFGKTVALPDHQTLSHRLPWAEGNREDGYLWGLDNMICLLQGLAQS | |
RUO | |
ITPKC | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
101.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IP3KC | |
ITPKC | |
Yes | |
wheat germ expression system |