Learn More
Abnova™ Human IMP4 Full-length ORF (NP_219484.1, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00092856-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
IMP4 forms a ternary complex with IMP3 (MIM 612980) and MPP10 (MPHOSPHO10; MIM 605503) that interacts with U3 small nucleolar RNA (snoRNA), which is required for the early cleavage steps in pre-rRNA processing (Granneman et al., 2003 [PubMed 12655004]).[supplied by OMIM]
Sequence: MLRREARLRREYLYRKAREEAQRSAQERKERLRRALEENRLIPTELRREALALQGSLEFDDAGGEGVTSHVDDEYRWAGVEDPKVMITTSRDPSSRLKMFAKELKLVFPGAQRMNRGRHEVGALVRACKANGVTDLLVVHEHRGTPVGLIVSHLPFGPTAYFTLCNVVMRHDIPDLGTMSEAKPHLITHGFSSRLGKRVSDILRYLFPVPKDDSHRVITFANQDDYISFRHHVYKKTDHRNVELTEVGPRFELKLYMIRLGTLEQEATADVEWRWHPYTNTARKRVFLSTEEspecificaciones
NP_219484.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BXDC4/MGC19606 | |
IMP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
92856 | |
IMP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLRREARLRREYLYRKAREEAQRSAQERKERLRRALEENRLIPTELRREALALQGSLEFDDAGGEGVTSHVDDEYRWAGVEDPKVMITTSRDPSSRLKMFAKELKLVFPGAQRMNRGRHEVGALVRACKANGVTDLLVVHEHRGTPVGLIVSHLPFGPTAYFTLCNVVMRHDIPDLGTMSEAKPHLITHGFSSRLGKRVSDILRYLFPVPKDDSHRVITFANQDDYISFRHHVYKKTDHRNVELTEVGPRFELKLYMIRLGTLEQEATADVEWRWHPYTNTARKRVFLSTE | |
RUO | |
IMP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |