Learn More
Abnova™ Human HSD17B8 Partial ORF (NP_055049, 28 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007923-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
In mice, the Ke6 protein is a 17-beta-hydroxysteroid dehydrogenase that can regulate the concentration of biologically active estrogens and androgens. It is preferentially an oxidative enzyme and inactivates estradiol, testosterone, and dihydrotestosterone. However, the enzyme has some reductive activity and can synthesize estradiol from estrone. The protein encoded by this gene is similar to Ke6 and is a member of the short-chain dehydrogenase superfamily. An alternatively spliced transcript of this gene has been detected, but the full-length nature of this variant has not been determined. [provided by RefSeq]
Sequence: SVRLAGEGATVAACDLDRAAAQETVRLLGGPGSKEGPPRGNHAAFQADVSEARAARCLLEQVQACFSRPPSVVVSCAGITQDEFLLHMSEDDWDKVIAVNEspecificaciones
NP_055049 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SVRLAGEGATVAACDLDRAAAQETVRLLGGPGSKEGPPRGNHAAFQADVSEARAARCLLEQVQACFSRPPSVVVSCAGITQDEFLLHMSEDDWDKVIAVN | |
RUO | |
HSD17B8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7923 | |
HSD17B8 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D6S2245E/FABG/FABGL/H2-KE6/HKE6/KE6/RING2/SDR30C1/dJ1033B10.9 | |
HSD17B8 | |
Recombinant | |
wheat germ expression system |