Learn More
Abnova™ Human HRMT1L6 Partial ORF (NP_060607, 241 a.a. - 316 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055170-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Protein arginine N-methyltransferases, such as PRMT6, catalyze the sequential transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine.[supplied by OMIM]
Sequence: ESEKPLVLSTSPFHPATHWKQALLYLNEPVQVEQDTDVSGEITLLPSRDNPRRLRVLLRYKVGDQEEKTKDFAMEDEspecificaciones
NP_060607 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ESEKPLVLSTSPFHPATHWKQALLYLNEPVQVEQDTDVSGEITLLPSRDNPRRLRVLLRYKVGDQEEKTKDFAMED | |
RUO | |
PRMT6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55170 | |
HRMT1L6 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10559/HRMT1L6 | |
PRMT6 | |
Recombinant | |
wheat germ expression system |