Learn More
Abnova™ Human HERC6 Partial ORF (NP_060382, 301 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055008-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
HERC6 belongs to the HERC family of ubiquitin ligases, all of which contain a HECT domain and at least 1 RCC1 (MIM 179710)-like domain (RLD). The 350-amino acid HECT domain is predicted to catalyze the formation of a thioester with ubiquitin before transferring it to a substrate, and the RLD is predicted to act as a guanine nucleotide exchange factor for small G proteins (Hochrainer et al., 2005 [PubMed 15676274]).[supplied by OMIM]
Sequence: AYVHTTGQVVSFGHGPSDTSKPTHPEALTENFDISCLISAEDFVDVQVKHIFAGTYANFVTTHQDTSSTRAPGKTLPEISRISQSMAEKWIAVKRRSTEHEspecificaciones
NP_060382 | |
Liquid | |
55008 | |
HERC6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20637 | |
HERC6 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AYVHTTGQVVSFGHGPSDTSKPTHPEALTENFDISCLISAEDFVDVQVKHIFAGTYANFVTTHQDTSSTRAPGKTLPEISRISQSMAEKWIAVKRRSTEH | |
RUO | |
HERC6 | |
Wheat Germ (in vitro) | |
GST |