Learn More
ACROBiosystems Human Her2 / ErbB2 (498-648) Protein, His Tag
423.38€ - 3108.82€
Especificaciones
Número de acceso | NP_004439.2 |
---|---|
Peso molecular | 18.5 kDa |
Fuente | Human Her2 (498-648), His Tag (HE2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 498 - Ala 648 (Accession # P04626-1). |
Inmunógeno | Asn 498 - Ala 648 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17186361
|
ACROBiosystems
HE2-H52H4-1MG |
1 mg |
3108.82€
1 miligramo |
Fecha estimada de envío: 08-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
17176361
|
ACROBiosystems
HE2-H52H4-100UG |
100 μg |
423.38€
100 microgramos |
Fecha estimada de envío: 08-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Human Epidermal growth factor Receptor 2 (HER2) is also called ERBB2, HER-2,HER-2 /neu, NEU, NGL,TKR1 and c-erb B2,and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family, more commonly known as the epidermal growth factor receptor family. HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor, with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product. Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development, cancer, communication at the neuromuscular junction and regulation of cell growth and differentiation .
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.5 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation. This protein has been verified not recognized by PertuEspecificaciones
NP_004439.2 | |
Human Her2 (498-648), His Tag (HE2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 498 - Ala 648 (Accession # P04626-1). | |
-20°C | |
Her2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
18.5 kDa | |
Asn 498 - Ala 648 | |
ERBB2,CD340,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |