Learn More
ACROBiosystems Human Her2 / ErbB2 (498-648) Protein, His Tag
Marca:  ACROBiosystems HE2-H52H4-1mg
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17186361
  • 3108.82€ / 1 miligramo
Fecha estimada de envío: 15-05-2024
para ver el stock
Descripción
Descripción
Human Epidermal growth factor Receptor 2 (HER2) is also called ERBB2, HER-2,HER-2 /neu, NEU, NGL,TKR1 and c-erb B2,and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family, more commonly known as the epidermal growth factor receptor family. HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor, with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product. Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development, cancer, communication at the neuromuscular junction and regulation of cell growth and differentiation .
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.5 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation. This protein has been verified not recognized by Pertu
Especificaciones
Especificaciones
NP_004439.2
1 mg
Asn 498 - Ala 648
ERBB2,CD340,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
18.5 kDa
Human Her2 (498-648), His Tag (HE2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 498 - Ala 648 (Accession # P04626-1).
-20°C
Her2
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens