Learn More
Abnova™ Human GOLGA1 Partial ORF (NP_002068, 668 a.a. - 767 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002800-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome. [provided by RefSeq]
Sequence: EKPGPEMANMAPSVTNNTDLTDAREINFEYLKHVVLKFMSCRESEAFHLIKAVSVLLNFSQEEENMLKETLEYKMSWFGSKPAPKGSIRPSISNPRIPWSEspecificaciones
NP_002068 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EKPGPEMANMAPSVTNNTDLTDAREINFEYLKHVVLKFMSCRESEAFHLIKAVSVLLNFSQEEENMLKETLEYKMSWFGSKPAPKGSIRPSISNPRIPWS | |
RUO | |
GOLGA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2800 | |
GOLGA1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC33154/golgin-97 | |
GOLGA1 | |
Recombinant | |
wheat germ expression system |