Learn More
Abnova™ Human GHRHR Partial ORF (NP_000814, 23 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002692-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene, expressed in the pituitary, encodes a receptor for growth hormone-releasing hormone. Binding of this hormone to the receptor leads to synthesis and release of growth hormone. Mutations in this gene have been associated with isolated growth hormone deficiency (IGHD), also known as Dwarfism of Sindh, a disorder characterized by short stature. Many alternate transcriptional splice variants encoding different isoforms have been described, but only two have been characterized to date. [provided by RefSeq]
Sequence: HMHPECDFITQLREDESACLQAAEEMPNTTLGCPATWDGLLCWPTAGSGEWVTLPCPDFFSHFSSESGAVKRDCTITGWSEPFPPYPVACPVPLELLAEEESYFSTVKIIEspecificaciones
NP_000814 | |
Liquid | |
2692 | |
GHRHR (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GHRFR/GHRHRpsv/GRFR | |
GHRHR | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HMHPECDFITQLREDESACLQAAEEMPNTTLGCPATWDGLLCWPTAGSGEWVTLPCPDFFSHFSSESGAVKRDCTITGWSEPFPPYPVACPVPLELLAEEESYFSTVKII | |
RUO | |
GHRHR | |
Wheat Germ (in vitro) | |
GST |