Learn More
Abnova™ Human FRG1 Partial ORF (NP_004468.1, 159 a.a. - 258 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002483-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene maps to a location 100 kb centromeric of the repeat units on chromosome 4q35 which are deleted in facioscapulohumeral muscular dystrophy (FSHD). It is evolutionarily conserved and has related sequences on multiple human chromosomes but DNA sequence analysis did not reveal any homology to known genes. In vivo studies demonstrate the encoded protein is localized to the nucleolus. [provided by RefSeq]
Sequence: CNEAGDIEAKSKTAGEEEMIKIRSCAERETKKKDDIPEEDKGNVKQCEINYVKKFQSFQDHKLKISKEDSKILKKARKDGFLHETLLDRRAKLKADRYCKEspecificaciones
NP_004468.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CNEAGDIEAKSKTAGEEEMIKIRSCAERETKKKDDIPEEDKGNVKQCEINYVKKFQSFQDHKLKISKEDSKILKKARKDGFLHETLLDRRAKLKADRYCK | |
RUO | |
FRG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2483 | |
FRG1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FRG1A/FSG1 | |
FRG1 | |
Recombinant | |
wheat germ expression system |