missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FPR2 Partial ORF (AAH29125.1, 163 a.a. - 205 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002358-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: FLTTVTIPNGDTYCTFNFASWGGTPEERLKVAITMLTARGIIREspecificaciones
AAH29125.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
30.36kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FLTTVTIPNGDTYCTFNFASWGGTPEERLKVAITMLTARGIIR | |
RUO | |
FPR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2358 | |
FPR2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALXR/FMLP-R-II/FMLPX/FPR2A/FPRH1/FPRH2/FPRL1/HM63/LXA4R | |
FPR2 | |
Recombinant | |
wheat germ expression system |