missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXG1B Partial ORF (NP_005240, 183 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002290-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16168174

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 28-04-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon. [provided by RefSeq]

Sequence: PFSYNALIMMAIRQSPEKRLTLNGIYEFIMKNFPYYRENKQGWQNSIRHNLSLNKCFVKVPRHYDDPGKGNYWMLDPSSDDVFIGGTTGKLRRRSTTSRAKLAFKRGAR
Especificaciones

Especificaciones

NP_005240
Liquid
2290
FOXG1B (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BF1/BF2/FHKL3/FKH2/FKHL1/FKHL2/FKHL3/FKHL4/FOXG1A/FOXG1B/FOXG1C/HBF-1/HBF-2/HBF-3/HBF-G2/HBF2/HFK1/HFK2/HFK3/KHL2/QIN
FOXG1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PFSYNALIMMAIRQSPEKRLTLNGIYEFIMKNFPYYRENKQGWQNSIRHNLSLNKCFVKVPRHYDDPGKGNYWMLDPSSDDVFIGGTTGKLRRRSTTSRAKLAFKRGAR
RUO
FOXG1
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales