Learn More
Abnova™ Human FOXG1 Partial ORF (CAA52241.1, 164 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002290-Q03.10ug
Detalles adicionales : Peso : 0.02000kg
Descripción
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon. [provided by RefSeq]
Sequence: SPFLSLHHPRASSTLSYNGTTSAYPSHPMPYSSVLTQNSLGNNHSFSTANGLSVDRLVNGEEspecificaciones
CAA52241.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.45kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
SPFLSLHHPRASSTLSYNGTTSAYPSHPMPYSSVLTQNSLGNNHSFSTANGLSVDRLVNGE | |
RUO | |
FOXG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2290 | |
FOXG1 (Human) Recombinant Protein (Q03) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BF1/BF2/FHKL3/FKH2/FKHL1/FKHL2/FKHL3/FKHL4/FOXG1A/FOXG1B/FOXG1C/HBF-1/HBF-2/HBF-3/HBF-G2/HBF2/HFK1/HFK2/HFK3/KHL2/QIN | |
FOXG1 | |
Recombinant | |
wheat germ expression system |