missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc epsilon RI alpha Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB6678
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Fc epsilon RI alpha Monoclonal specifically detects Fc epsilon RI alpha in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.
Especificaciones
Fc epsilon RI alpha | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.08-4 ug/mL, CyTOF-ready | |
P12319 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2205 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, Neutralization, CyTOF | |
773704 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FCER1A | |
Mouse myeloma cell line NS0-derived recombinant human Fc epsilon RI alpha Val26-Gln205 Accession # P12319 | |
100 μg | |
Primary | |
Detects human Fc epsilon RI alpha in direct ELISAs. | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human Fc epsilon RI alpha Antibody, R&D Systems™ > 100ug; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido