missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO36 Full-length ORF (AAH33935.1, 1 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00130888-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16107913

  • 518.16€ / 25 microgramos
Envío estimado: 08-03-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Members of the F-box protein family, such as FBXO36, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MASWLPETLFETVGQGPPPSKDYYQLLVTRSQVIFRWWKISLRSEYRSTKPGEAKETHEDFLENSHLQGQTALIFGARILDYVINFCKGKFDFLERLSDDLLLTIISYLDLEDIARLCQTSHRFAKLCMSDKLWEQIVQSTCDTITPDVRALAEDTGWRQLFFTNKLQLQRQLRKRKQKYGNLREKQP
Especificaciones

Especificaciones

AAH33935.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ37592/FLJ41090/Fbx36
FBXO36
Yes
wheat germ expression system
Liquid
130888
FBXO36 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASWLPETLFETVGQGPPPSKDYYQLLVTRSQVIFRWWKISLRSEYRSTKPGEAKETHEDFLENSHLQGQTALIFGARILDYVINFCKGKFDFLERLSDDLLLTIISYLDLEDIARLCQTSHRFAKLCMSDKLWEQIVQSTCDTITPDVRALAEDTGWRQLFFTNKLQLQRQLRKRKQKYGNLREKQP
RUO
FBXO36
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales