Learn More
Abnova™ Human ELP4 Full-length ORF (AAH12514.1, 1 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00026610-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined. [provided by RefSeq]
Sequence: MAAVATCGSVAASTGSAVATASKSNVTSFQRRGPRASVTNDSGPRLVSIAGTRPSVRNGQLLVSTGLPALDQLLGGGLAVGTVLLIEEDKYNIYSPLLFKYFLAEGIVNGHTLLVASAKEDPANILQELPAPLLDDKCKKEFDEDVYNHKTPESNIKMKIAWRYQLLPKMEIGPVSSSRFGHYYDASKRMPQELIEASNWHGFFLPEKISSTLKVEPCSLTPGYTKLLQFIQNIIYEEGFDGSNPQKKQRNILRIGIQNLGSPLWGDDICCAENGGNSHSLTKFLYVLRGLLRTSLSACIITMPTHLIQNKAIIARVTTLSDVVVGLESFIGSERETNPLYKDYHGLIHIRQIPRLNNLICDESDVKDLAFKLKRKLFTIEAGVQWHDLGSRRPRLLGSGGSPASASLVAGITGAHHHAQLIFVFLVEMGFHHVGQAGLELLTSGDSSASASQSAGIAGMSYRARPRALYFKENKSKVGARQLLETREEHLSSRLLILTQAERLCMGRRFFTAFHIFNELPCKGDCICLQTCQTQEspecificaciones
AAH12514.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
85.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C11orf19/FLJ20498/PAX6NEB/PAXNEB/dJ68P15A.1 | |
ELP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26610 | |
ELP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAVATCGSVAASTGSAVATASKSNVTSFQRRGPRASVTNDSGPRLVSIAGTRPSVRNGQLLVSTGLPALDQLLGGGLAVGTVLLIEEDKYNIYSPLLFKYFLAEGIVNGHTLLVASAKEDPANILQELPAPLLDDKCKKEFDEDVYNHKTPESNIKMKIAWRYQLLPKMEIGPVSSSRFGHYYDASKRMPQELIEASNWHGFFLPEKISSTLKVEPCSLTPGYTKLLQFIQNIIYEEGFDGSNPQKKQRNILRIGIQNLGSPLWGDDICCAENGGNSHSLTKFLYVLRGLLRTSLSACIITMPTHLIQNKAIIARVTTLSDVVVGLESFIGSERETNPLYKDYHGLIHIRQIPRLNNLICDESDVKDLAFKLKRKLFTIEAGVQWHDLGSRRPRLLGSGGSPASASLVAGITGAHHHAQLIFVFLVEMGFHHVGQAGLELLTSGDSSASASQSAGIAGMSYRARPRALYFKENKSKVGARQLLETREEHLSSRLLILTQAERLCMGRRFFTAFHIFNELPCKGDCICLQTCQTQ | |
RUO | |
ELP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |