missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EDNRB/Endothelin R Type B Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB4496
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
EDNRB/Endothelin R Type B Monoclonal specifically detects EDNRB/Endothelin R Type B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Especificaciones
EDNRB/Endothelin R Type B | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-25 ug/mL, CyTOF-ready | |
P24530 | |
EDNRB | |
NS0 mouse myeloma cell line transfected with human EDNRB/Endothelin R Type B Accession # P24530 | |
100 μg | |
Primary | |
Detects human EDNRB/Endothelin R Type B in direct ELISAs. | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
671917 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ABCDS, EDNRB, endothelin B receptor, Endothelin receptor non-selective type, endothelin receptor type B, ETB, ET-B, ETBR, ET-BR, ETRB, HSCR, HSCR2, WS4A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1910 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human EDNRB/Endothelin R Type B Antibody, R&D Systems™ > 100ug; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido