Learn More
Abnova™ Human COG4 Partial ORF (NP_056201, 686 a.a. - 785 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00025839-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Multiprotein complexes are key determinants of Golgi apparatus structure and its capacity for intracellular transport and glycoprotein modification. Several complexes have been identified, including the Golgi transport complex (GTC), the LDLC complex, which is involved in glycosylation reactions, and the SEC34 complex, which is involved in vesicular transport. These 3 complexes are identical and have been termed the conserved oligomeric Golgi (COG) complex, which includes COG4 (Ungar et al., 2002 [PubMed 11980916]).[supplied by OMIM]
Sequence: ELEKVVLKSTFNRLGGLQFDKELRSLIAYLTTVTTWTIRDKFARLSQMATILNLERVTEILDYWGPNSGPLTWRLTPAEVRQVLALRIDFRSEDIKRLRLEspecificaciones
NP_056201 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ELEKVVLKSTFNRLGGLQFDKELRSLIAYLTTVTTWTIRDKFARLSQMATILNLERVTEILDYWGPNSGPLTWRLTPAEVRQVLALRIDFRSEDIKRLRL | |
RUO | |
COG4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25839 | |
COG4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COD1/DKFZp586E1519 | |
COG4 | |
Recombinant | |
wheat germ expression system |