Learn More
Abnova™ Human CLN6 Full-length ORF (NP_060352.1, 1 a.a. - 311 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054982-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is one of eight which have been associated with neuronal ceroid lipofuscinoses (NCL). Also referred to as Batten disease, NCL comprises a class of autosomal recessive, neurodegenerative disorders affecting children. The genes responsible likely encode proteins involved in the degradation of post-translationally modified proteins in lysosomes. The primary defect in NCL disorders is thought to be associated with lysosomal storage function. [provided by RefSeq]
Sequence: MEATRRRQHLGATGGPGAQLGASFLQARHGSVSADEAARTAPFHLDLWFYFTLQNWVLDFGRPIAMLVFPLEWFPLNKPSVGDYFHMAYNVITPFLLLKLIERSPRTLPRSITYVSIIIFIMGASIHLVGDSVNHRLLFSGYQHHLSVRENPIIKNLKPETLIDSFELLYYYDEYLGHCMWYIPFFLILFMYFSGCFTASKAESLIPGPALLLVAPSGLYYWYLVTEGQIFILFIFTFFAMLALVLHQKRKRLFLDSNGLFLFSSFALTLLLVALWVAWLWNDPVLRKKYPGVIYVPEPWAFYTLHVSSRHEspecificaciones
NP_060352.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20561/HsT18960/nclf | |
CLN6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54982 | |
CLN6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEATRRRQHLGATGGPGAQLGASFLQARHGSVSADEAARTAPFHLDLWFYFTLQNWVLDFGRPIAMLVFPLEWFPLNKPSVGDYFHMAYNVITPFLLLKLIERSPRTLPRSITYVSIIIFIMGASIHLVGDSVNHRLLFSGYQHHLSVRENPIIKNLKPETLIDSFELLYYYDEYLGHCMWYIPFFLILFMYFSGCFTASKAESLIPGPALLLVAPSGLYYWYLVTEGQIFILFIFTFFAMLALVLHQKRKRLFLDSNGLFLFSSFALTLLLVALWVAWLWNDPVLRKKYPGVIYVPEPWAFYTLHVSSRH | |
RUO | |
CLN6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |