Learn More
Abnova™ Human CDC37L1 Full-length ORF (AAH14133, 1 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055664-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 (HSPCA; MIM 140571) as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 [PubMed 11413142]).[supplied by OMIM]
Sequence: MEQPWPPPGPWSLPRAEGEAEEESDFDVFPSSPRCPQLPGGGAQMYSHGIELACQKQKEFVKSSVACKWNLAEAQQKLGSLALHNSESLDQEHAKAQTAVSELRQREEEWRQKEEALVQREKMCLWSTDAISKDVFNKSFINQDKRKDTEDEDKSESFMQKYEQKIRHFGMLSRWDDSQRFLSDHPYLVCEETAKYLILWCFHLEAEKKGALMEQIAHQAVVMQFIMEMAKNCNVDPRGCFRLFFQKAKAEEEGYFEAFKNELEAFKSRVRLYSQSQSFQPMTVQNHVPHSGVGSIGLLESLPQNPDYLQYSISTALCSLNSVVHKEDDEPKMMDTVEspecificaciones
AAH14133 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.81kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDC37B/FLJ20639/HARC | |
CDC37L1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55664 | |
CDC37L1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEQPWPPPGPWSLPRAEGEAEEESDFDVFPSSPRCPQLPGGGAQMYSHGIELACQKQKEFVKSSVACKWNLAEAQQKLGSLALHNSESLDQEHAKAQTAVSELRQREEEWRQKEEALVQREKMCLWSTDAISKDVFNKSFINQDKRKDTEDEDKSESFMQKYEQKIRHFGMLSRWDDSQRFLSDHPYLVCEETAKYLILWCFHLEAEKKGALMEQIAHQAVVMQFIMEMAKNCNVDPRGCFRLFFQKAKAEEEGYFEAFKNELEAFKSRVRLYSQSQSFQPMTVQNHVPHSGVGSIGLLESLPQNPDYLQYSISTALCSLNSVVHKEDDEPKMMDTV | |
RUO | |
CDC37L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |