Learn More
Abnova™ Human CDC14A Partial ORF (NP_003663, 431 a.a. - 530 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008556-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. It is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, suggesting a role in cell cycle control. This protein has been shown to interact with, and dephosphorylate tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splicing of this gene results in several transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: PFRLSSSLQGSAVTLKTSKMALSPSATAKRINRTSLSSGATVRSFSINSRLASSLGNLNAATDDPENKKTSSSSKAGFTASPFTNLLNGSSQPTTRNYPEEspecificaciones
NP_003663 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PFRLSSSLQGSAVTLKTSKMALSPSATAKRINRTSLSSGATVRSFSINSRLASSLGNLNAATDDPENKKTSSSSKAGFTASPFTNLLNGSSQPTTRNYPE | |
RUO | |
CDC14A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8556 | |
CDC14A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
cdc14/hCDC14 | |
CDC14A | |
Recombinant | |
wheat germ expression system |