Learn More
Abnova™ Human CDAN1 Partial ORF (NP_612486.2, 1130 a.a. - 1227 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00146059-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that appears to play a role in nuclear envelope integrity, possibly related to microtubule attachments. Mutations in this gene cause congenital dyserythropoietic anemia type I, a disease resulting in morphological and functional abnormalities of erythropoiesis. [provided by RefSeq]
Sequence: PLQLLLSPRNVGLLADTRPREWDLLLFLLRELVEKGLMGRMEIEACLGSLHQAQWPGDFAEELATLSNLFLAEPHLPEPQLRACELVQPNRGTVLAQSEspecificaciones
NP_612486.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PLQLLLSPRNVGLLADTRPREWDLLLFLLRELVEKGLMGRMEIEACLGSLHQAQWPGDFAEELATLSNLFLAEPHLPEPQLRACELVQPNRGTVLAQS | |
RUO | |
CDAN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
146059 | |
CDAN1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDA-I/CDA1/CDAI/DLT/PRO1295/codanin | |
CDAN1 | |
Recombinant | |
wheat germ expression system |