missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin D Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB10142-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cathepsin D Monoclonal specifically detects Cathepsin D in Human samples. It is validated for ELISA.
Especificaciones
Cathepsin D | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
Mouse myeloma cell line, NS0-derived human Cathepsin D protein, Leu21-Leu412, Accession No. P07339.1 | |
100 μg | |
1509.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
ELISA | |
185109 | |
ELISA | |
P07339.1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Cathepsin D in direct ELISAs. | |
Human | |
Supernatant |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human Cathepsin D Antibody, R&D Systems™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido